Merck
  • Guanidinylation of Chitooligosaccharides Involving Internal Cyclization of the Guanidino Group on the Reducing End and Effect of Guanidinylation on Protein Binding Ability.
Guanidinylation of Chitooligosaccharides Involving Internal Cyclization of the Guanidino Group on the Reducing End and Effect of Guanidinylation on Protein Binding Ability.
Biomolecules (2019-07-10)
Hironori Izawa, Mizuki Kinai, Shinsuke Ifuku, Minoru Morimoto, Hiroyuki Saimoto
RESUMO
In order to synthesize a promising material for developing a novel peptide/protein delivery system, guanidinylation of chitooligosaccharides with 1-amidinopyrazole hydrochloride was investigated herein. The production of guanidinylated chitooligosaccharides was demonstrated by infrared spectroscopy (IR), nuclear magnetic resonance (NMR), and elemental analyses. Interestingly, we found that the reducing end in the guanidinylated chitooligosaccharides was converted to a cyclic guanidine structure (2-[(aminoiminomethyl)amino]-2-deoxy-d-glucose structure). This reaction was carefully proven by the guanidinylation of d-glucosamine. Although this is not the first report on the synthesis of the 2-[(aminoiminomethyl)amino]-2-deoxy-d-glucose, it has provided a rational synthetic route using the high reactivity of the reducing end. Furthermore, we found that the interaction between chitooligosaccharides and bovine serum albumin is weak when in a neutral pH environment; however, it is significantly improved by guanidinylation. The guanidinylated chitooligosaccharides are useful not only for the development of a novel drug delivery system but also as a chitinase/chitosanase inhibitor and an antibacterial agent.