Merck
  • Novel insights in biopurification system for dissipation of a pesticide mixture in repeated applications.
Novel insights in biopurification system for dissipation of a pesticide mixture in repeated applications.
Environmental science and pollution research international (2017-09-16)
Maria Cristina Diez, Bárbara Leiva, Felipe Gallardo
RESUMO
A biopurification system based on the adsorption and degradation capacity of a biomixture to degrade a mixture of pesticides (atrazine, chlorpyrifos, iprodione; 50 mg kg-1 each) in repeated applications (0, 30, and 60 days) was evaluated. Tanks of 1 m3 packed with a biomixture (ρ 0.29 g mL-1) with and without vegetal cover were used. The biomixture contained soil, peat, and wheat straw in a proportion 1:1:2 by volume, respectively. Pesticide concentrations, biological activities (urease, phenoloxidase, and dehydrogenase), and microbial community changes (DGGE and qPCR) were evaluated periodically. Pesticide dissipation was higher in tanks with vegetal cover (> 95%) and no variation was observed after the three applications; contrarily, pesticide dissipation decreased in the tank without vegetal cover after each application. The presence of vegetal cover decreased the half-life of pesticides by at least twice. Biological activities were in general not affected by the application and reapplication of pesticides in the same treatment; however, they exhibited some differences between tanks containing and lacking the vegetal cover. High similarity between microbial groups (actinobacteria, bacteria, and fungi) was observed, suggesting no influence ascribable to the successive pesticide applications. The number of copies of bacteria and actinobacteria remained almost constant during the assay. However, the number of copies of fungi was significantly higher in the uncontaminated tank without vegetal cover.