Merck
Binary tree-inspired digital dendrimer.
Nature communications (2019-04-25)
Zhihao Huang, Qiunan Shi, Jiang Guo, Fanying Meng, Yajie Zhang, Yutong Lu, Zhuangfei Qian, Xiaopeng Li, Nianchen Zhou, Zhengbiao Zhang, Xiulin Zhu
RESUMO
Digital polymers with precisely ordered units acting as the coded 0- or 1-bit, are introduced as a promising option for molecular data storage. However, the pursuit of better performance in terms of high storage capacity and useful functions never stops. Herein, we propose a concept of an information-coded 2D digital dendrimer. The divergent growth via thiol-maleimide Michael coupling allows precise arrangements of the 0- and 1-bits in the uniform dendrimers. A protocol for calculating the storage capacity of non-linear binary digital dendrimer is established based on data matrix barcode, generated by the tandem mass spectrometry decoding and encryption. Furthermore, the generated data matrix barcode can be read by a common hand-held device to cater the applications such as item identification, traceability and anticouterfeiting purpose. This work demonstrates the high data storage capacity of a uniform dendrimer and uncovers good opportunities for the digital polymers.