Merck
Targeted metabolomics of whole blood using volumetric absorptive microsampling.
Talanta (2019-02-18)
Miranda G M Kok, Cindy Nix, Gwenaël Nys, Marianne Fillet
RESUMO
Volumetric absorptive microsampling (VAMS) enables the collection of small and accurate quantities of biological fluids. Therefore, this sampling technique is of great interest for volume-limited samples or serial collection of samples. In this study, we examined the potential of VAMS for targeted mass spectrometry (MS)-based metabolomics. The targeted analysis of 36 major metabolites from only 10 μL of whole blood was optimized. A design of experiments was carried out to maximize the extraction of metabolites. Moreover, critical steps in sample preparation and sample analysis were studied and characterized, such as the addition of internal standards to tips of VAMS devices before sample collection. A reversed-phase UHPLC-MS/MS method was used to analyze organic acids, whereas hydrophilic interaction chromatography (HILIC)-MS/MS was selected for the determination of amino acids. Overall, the optimum extraction solvent was acetonitrile-water in a proportion of 60:40 (v/v), providing good recoveries and resulting in the detection of all target metabolites in whole blood with good repeatability (less than 15% RSD on peak area). Furthermore, the stability of the analytes in dried whole blood, which is of critical importance in metabolomics studies, was investigated. The amino and organic acids were stable for at least 4 days when stored at room temperature. This is in contrast to the instability of these compounds in wet blood, thereby showing the great potential of VAMS in metabolomics studies.