Merck
Plasma syndecan-1 in hemodialysis patients associates with survival and lower markers of volume status.
American journal of physiology. Renal physiology (2018-11-01)
Josephine Koch, Nienke M A Idzerda, Wendy Dam, Solmaz Assa, Casper F M Franssen, Jacob van den Born
RESUMO
Syndecan-1, a transmembrane heparan sulfate proteoglycan, associates with renal and cardiovascular functioning. We earlier reported syndecan-1 to be involved in renal tubular regeneration. We now examined plasma values of syndecan-1 in a hemodialysis cohort and its association with volume and inflammatory and endothelial markers in addition to outcome. Eighty-four prevalent hemodialysis patients were evaluated for their plasma syndecan-1 levels by ELISA before the start of hemodialysis, as well as 60, 180, and 240 min after start of dialysis. Patients were divided into sex-stratified tertiles based on predialysis plasma syndecan-1 levels. We studied the association between plasma levels of syndecan-1 and volume, inflammation, and endothelial markers and its association with cardiovascular events and all-cause mortality using Kaplan-Meier curves and Cox regression analyses with adjustments for gender, age, diabetes, and dialysis vintage. Predialysis syndecan-1 levels were twofold higher in men compared with women ( P = 0.0003). Patients in the highest predialysis plasma syndecan-1 tertile had a significantly higher ultrafiltration rate ( P = 0.034) and lower plasma values of BNP ( P = 0.019), pro-ANP ( P = 0.024), and endothelin ( P < 0.0001) compared with the two lower predialysis syndecan-1 tertiles. No significant associations with inflammatory markers were found. Cox regression analysis showed that patients in the highest syndecan-1 tertile had significantly less cardiovascular events and better survival compared with the lowest syndecan-1 tertile ( P = 0.02 and P = 0.005, respectively). In hemodialysis patients, higher plasma syndecan-1 levels were associated with lower concentrations of BNP, pro-ANP, and endothelin and with better patient survival. This may suggest that control of volume status in hemodialysis patients allows an adaptive tissue regenerative response as reflected by higher plasma syndecan-1 levels.