Merck
  • The long intergenic non-protein coding RNA 707 promotes proliferation and metastasis of gastric cancer by interacting with mRNA stabilizing protein HuR.
The long intergenic non-protein coding RNA 707 promotes proliferation and metastasis of gastric cancer by interacting with mRNA stabilizing protein HuR.
Cancer letters (2018-12-07)
Min Xie, Tianshi Ma, Jiangyang Xue, Hongwei Ma, Ming Sun, Zhihong Zhang, Minjuan Liu, Yinghua Liu, Songwen Ju, Zhaoxia Wang, Wei De
RESUMO
Multiple studies have revealed that long non-coding RNAs (lncRNAs) extensively participate in human cancer malignant progression. The long intergenic non-protein coding RNA 707 (LINC00707), 3087 bp in length, was recently reported to be an essential oncogene in promoting lung adenocarcinoma cell proliferation and metastasis. However, its role in gastric cancer (GC) remains unclear. In this study, we identified that LINC00707 was excessively expressed in GC tissues and correlated with advanced stage, larger tumor size, lymph node metastasis and poorer prognosis in GC patients. In vitro and in vivo assays showed that LINC00707 promote GC cell proliferation and metastasis. Mechanistically, LINC00707 could abundantly interact with mRNA stabilizing protein HuR; "LINC00707-HuR" coalition ulteriorly combined with VAV3/F11R mRNAs and increased their stability. Taken together, our findings prove that LINC00707 may act as an oncogene in GC by regulating mRNA stability and serve as a potential target for GC diagnosis and prognosis.