Merck
Development of a high-throughput fluorescent no-wash sodium influx assay.
PloS one (2019-03-12)
Bryan Tay, Teneale A Stewart, Felicity M Davis, Jennifer R Deuis, Irina Vetter
RESUMO
Voltage-gated sodium channels (NaVs) are key therapeutic targets for pain, epilepsy and cardiac arrhythmias. Here we describe the development of a no-wash fluorescent sodium influx assay suitable for high-throughput screening and characterization of novel drug leads. Addition of red-violet food dyes (peak absorbance range 495-575 nm) to assays in HEK293 cells heterologously expressing hNaV1.1-1.8 effectively quenched background fluorescence of the sodium indicator dye Asante NaTRIUM Green-2 (ANG-2; peak emission 540 nm), negating the need for a wash step. Ponceau 4R (1 mM) was identified as a suitable quencher, which had no direct effect on NaV channels as assessed by patch-clamp experiments, and did not alter the pharmacology of the NaV1.1-1.7 activator veratridine (EC50 10-29 μM) or the NaV1.1-1.8 inhibitor tetracaine (IC50's 6-66 μM). In addition, we also identified that the food dyes Ponceau 4R, Brilliant Black BN, Allura Red and Amaranth are effective at quenching the background fluorescence of the calcium indicator dyes fluo-4, fura-2 and fura-5F, identifying them as potential inexpensive alternatives to no-wash calcium ion indicator kits. In summary, we have developed a no-wash fluorescent sodium influx assay suitable for high-throughput screening based on the sodium indicator dye ANG-2 and the quencher Ponceau 4R.