Merck
  • Macrophage-derived CCL22 promotes an immunosuppressive tumor microenvironment via IL-8 in malignant pleural effusion.
Macrophage-derived CCL22 promotes an immunosuppressive tumor microenvironment via IL-8 in malignant pleural effusion.
Cancer letters (2019-04-01)
Dong Wang, Li Yang, Dongli Yue, Ling Cao, Lifeng Li, Dan Wang, Yu Ping, Zhibo Shen, Yujia Zheng, Liping Wang, Yi Zhang
RESUMO
Immune dysfunction often occurs in malignant pleural effusion (MPE). In our previous study, TGF-β derived predominantly from macrophages plays an important role in impairing T cell cytotoxicity in MPE. Therefore, we aimed to investigate whether other immunoregulatory cells and factors mediated TGF-β secretion from macrophages, involved in the immunosuppressive microenvironment of MPE, and to provide clues for potential immune therapy for MPE as well. We found that CCL22 level in MPE was significantly higher than that in non-malignant pleural effusion. The high level of CCL22 was closely associated with poor survival in MPE patients with lung cancer. CCL22 was dominantly produced by tumor-associated macrophages (TAMs) in MPE. Meanwhile, TAM-derived TGF-β mediated CCL22 expression in TAMs via c-Fos. CCL22 promoted the recruitment of regulatory T cells (Tregs) in MPE. Lastly, Treg-secreted high level of IL-8 further induced TGF-β production from TAMs, and promoted the immunosuppressive tumor microenvironment in MPE. Our results indicate that macrophage-derived CCL22 plays an important role in the immunosuppressive tumor microenvironment via IL-8 in MPE.