Merck
  • Clopidogrel but Not Prasugrel Significantly Inhibits the CYP2C8-Mediated Metabolism of Montelukast in Humans.
Clopidogrel but Not Prasugrel Significantly Inhibits the CYP2C8-Mediated Metabolism of Montelukast in Humans.
Clinical pharmacology and therapeutics (2017-11-25)
Matti K Itkonen, Aleksi Tornio, Anne M Filppula, Mikko Neuvonen, Pertti J Neuvonen, Mikko Niemi, Janne T Backman
RESUMO
The oxidation of montelukast is mainly mediated by cytochrome P450 (CYP) 2C8, but other mechanisms may contribute to its disposition. In healthy volunteers, we investigated the effects of two widely used P2Y12 inhibitors on montelukast pharmacokinetics. Clopidogrel (300 mg on day 1 and 75 mg on day 2) increased the area under the plasma concentration-time curve (AUC) of montelukast 2.0-fold (90% confidence interval (CI) 1.72-2.28, P < 0.001) and decreased the M6:montelukast AUC0-7h ratio to 45% of control (90% CI 40-50%, P < 0.001). Prasugrel (60 mg on day 1 and 10 mg on day 2) had no clinically meaningful effect on montelukast pharmacokinetics. Our results imply that clopidogrel is at least a moderate inhibitor of CYP2C8, but prasugrel is not a clinically relevant CYP2C8 inhibitor. The different interaction potentials of clopidogrel and prasugrel are important to consider when antiplatelet therapy is planned for patients at risk for polypharmacy with CYP2C8 substrates.