Merck
  • Position of acetyl groups on anhydroglucose unit in acetylated starches with intermediate degrees of substitution.
Position of acetyl groups on anhydroglucose unit in acetylated starches with intermediate degrees of substitution.
Carbohydrate polymers (2019-06-15)
Jianteng Xu, Yong-Cheng Shi
RESUMO
Waxy, normal, and high-amylose maize starches were acetylated to various degrees of substitution (DS) in aqueous medium. A stepwise addition of acetic anhydride enhanced reaction efficiency and DS was in the order of high-amylose maize starch (DS 1.72) > waxy maize starch (DS 1.60) > normal maize starch (DS 1.03) when 240% (w/w) acetic anhydride was added. Starch acetates were further fully esterified to DS 3.0 using acetic anhydride-d6 or propionic anhydride to analyze position of acetyl groups on anhydroglucose unit (AGU) by 1H NMR. Regioselective 2-O-acetyl starches were produced when the DS was lower than 0.14. At DS 1, the distribution of acetyl groups was ca. 44%, 32%, and 24% at C-2, C-3, and C-6 positions, respectively, regardless of amylose content. Deuterated acetyl group was a preferred secondary substituent over propionyl group in subsequent 1H NMR analysis because it resulted a clean spectrum without interference from propionyl methyl protons.