Merck
UVA irradiation strengthened an interaction between UBF1/2 proteins and H4K20 di-/tri-methylation.
Chromosome research : an international journal on the molecular, supramolecular and evolutionary aspects of chromosome biology (2019-01-06)
Lenka Stixová, Denisa Komůrková, Alena Svobodová Kovaříková, Eva Bártová
RESUMO
Repair of ribosomal DNA (rDNA) is a very important nuclear process due to the most active transcription of ribosomal genes. Proper repair of rDNA is required for physiological biogenesis of ribosomes. Here, we analyzed the epigenetics of the DNA damage response in a nucleolar compartment, thus in the ribosomal genes studied in nonirradiated and UVA-irradiated mouse embryonic fibroblasts (MEFs). We found that the promoter of ribosomal genes is not abundant on H4K20me2, but it is densely occupied by H4K20me3. Ribosomal genes, regulated via UBF1/2 proteins, were characterized by an interaction between UBF1/2 and H4K20me2/me3. This interaction was strengthened by UVA irradiation that additionally causes a focal accumulation of H4K20me3 in the nucleolus. No interaction has been found between UBF1/2 and H3K9me3. Interestingly, UVA irradiation decreases the levels of H3K9me3 and H4K20me3 at 28S rDNA. Altogether, the UVA light affects the epigenetic status of ribosomal genes at 28S rDNA and strengthens an interaction between UBF1/2 proteins and H4K20me2/me3.