Merck
  • Co-Polymers based on Poly(1,4-butylene 2,5-furandicarboxylate) and Poly(propylene oxide) with Tuneable Thermal Properties: Synthesis and Characterization.
Co-Polymers based on Poly(1,4-butylene 2,5-furandicarboxylate) and Poly(propylene oxide) with Tuneable Thermal Properties: Synthesis and Characterization.
Materials (Basel, Switzerland) (2019-01-24)
Marina Matos, Andreia F Sousa, Patrícia V Mendonça, Armando J D Silvestre
RESUMO
Poly(ether ester)s (PEEs) represent a promising class of segmented co-polymers, nevertheless the synthesis of PEEs based on renewable 2,5-furandicarboxylic acid (FDCA) is still scarce. In this context, a series of poly(1,4-butylene 2,5-furandicarboxylate)-co-poly(poly(propylene oxide) 2,5-furandicarboxylate) co-polyesters with different composition of stiff poly(1,4-butylene 2,5-furandicarboxylate) (PBF) and soft poly(poly(propylene oxide) 2,5-furandicarboxylate) (PPOF) moieties were synthesized, via a two-step bulk polytransesterification reaction. The molar ratio of PBF/PPOF incorporated was varied (10 to 50 mol%) in order to prepare several novel materials with tuned properties. The materials were characterised in detail through several techniques, namely ATR FTIR, ¹H and 13C NMR, TGA, DSC, DMTA and XRD. Their hydrolytic and enzymatic degradation evaluation was also assessed. These new co-polymers showed either a semi-crystalline nature when higher PBF/PPOF ratios were used, and for approximately equal amounts of PBF and PPOF an amorphous co-polyester was obtained instead.