Merck
  • The AKT kinase signaling network is rewired by PTEN to control proximal BCR signaling in germinal center B cells.
The AKT kinase signaling network is rewired by PTEN to control proximal BCR signaling in germinal center B cells.
Nature immunology (2019-04-24)
Wei Luo, William Hawse, Laura Conter, Nikita Trivedi, Florian Weisel, Daniel Wikenheiser, Richard T Cattley, Mark J Shlomchik
RESUMO
B cell antigen receptor (BCR) and CD40 signaling are rewired in germinal center (GC) B cells (GCBCs) to optimize selection for high-affinity B cells. In GCBC, BCR signals are constrained, but the mechanisms are not well understood. Here we describe a GC-specific, AKT-kinase-driven negative feedback loop that attenuates BCR signaling. Mass spectrometry revealed that AKT target activity was altered in GCBCs compared with naive B cells. Retargeting was linked to differential AKT T308 and S473 phosphorylation, in turn controlled by GC-specific upregulation of phosphoinositide-dependent protein kinase PDK1 and the phosphatase PTEN. In GCBCs, AKT preferentially targeted CSK, SHP-1 and HPK1, which are negative regulators of BCR signaling. We found that phosphorylation enhances enzymatic activity of these proteins, creating a negative feedback loop that dampens upstream BCR signaling. AKT inhibition relieved this negative feedback and enhanced activation of BCR-proximal kinase LYN, as well as downstream BCR signaling molecules in GCBCs.