Merck
Leucine zipper-EF-hand containing transmembrane protein 1 (LETM1) forms a Ca2+/H+ antiporter.
Scientific reports (2016-09-28)
Juan Shao, Zhenglin Fu, Yanli Ji, Xiangchen Guan, Shang Guo, Zhanyu Ding, Xue Yang, Yao Cong, Yuequan Shen
RESUMO
Leucine zipper-EF-hand-containing transmembrane protein1 (LETM1) is located in the mitochondrial inner membrane and is defective in Wolf-Hirschhorn syndrome. LETM1 contains only one transmembrane helix, but it behaves as a putative transporter. Our data shows that LETM1 knockdown or overexpression robustly increases or decreases mitochondrial Ca2+ level in HeLa cells, respectively. Also the residue Glu221 of mouse LETM1 is identified to be necessary for Ca2+ flux. The mutation of Glu221 to glutamine abolishes the Ca2+-transport activity of LETM1 in cells. Furthermore, the purified LETM1 exhibits Ca2+/H+ anti-transport activity, and the activity is enhanced as the proton gradient is increased. More importantly, electron microscopy studies reveal a hexameric LETM1 with a central cavity, and also, observe two different conformational states under alkaline and acidic conditions, respectively. Our results indicate that LETM1 is a Ca2+/H+ antiporter and most likely responsible for mitochondrial Ca2+ output.