- Publisher Correction: Structural basis of ligand recognition at the human MT1 melatonin receptor.
Publisher Correction: Structural basis of ligand recognition at the human MT1 melatonin receptor.
Nature (2019-05-03)
Benjamin Stauch, Linda C Johansson, John D McCorvy, Nilkanth Patel, Gye Won Han, Xi-Ping Huang, Cornelius Gati, Alexander Batyuk, Samuel T Slocum, Andrii Ishchenko, Wolfgang Brehm, Thomas A White, Nairie Michaelian, Caleb Madsen, Lan Zhu, Thomas D Grant, Jessica M Grandner, Anna Shiriaeva, Reid H J Olsen, Alexandra R Tribo, Saïd Yous, Raymond C Stevens, Uwe Weierstall, Vsevolod Katritch, Bryan L Roth, Wei Liu, Vadim Cherezov
PMID31048811
RESUMO
Change history: In this Letter, the rotation signs around 90°, 135° and 15° were missing and in the HTML, Extended Data Tables 2 and 3 were the wrong tables; these errors have been corrected online.
MATERIAIS