Merck
  • Susceptibilities of phospholipid vesicles containing different sterols to amphotericin B-loaded lysophosphatidylcholine micelles.
Susceptibilities of phospholipid vesicles containing different sterols to amphotericin B-loaded lysophosphatidylcholine micelles.
Journal of biochemistry (2003-08-29)
Maki Onda, Yukari Inoue, Manami Kawabata, Tomoyoshi Mita
RESUMO
To investigate the susceptibilities of fungal and mammalian cells to amphotericin B (AmB), AmB-loaded lysophosphatidylcholine (LPC)micelles as drug delivery vehicles were incubated at 37 degrees C with phosphatidylcholine vesicles containing different sterols as model systems for fungal and mammalian cells. The binding and kinetics of AmB to sterols in the membranes were judged by UV-visible spectroscopy. In the 91% monomeric form, AmB interacted rapidly with ergosterol and slowly with 7-dehydrocholesterol (7-DHC), while it did not interact with cholesterol. In the 50% monomeric form, AmB formed complexes more rapidly with ergosterol or 7-DHC than in the monomeric form, whereas it did not still interact with cholesterol. The interaction was also characterized by resonance energy transfer between the fluorescent probe trimethylammonium diphenylhexatriene (TMA-DPH) and AmB. In the 91% monomeric form, AmB caused initial fluorescence quenching in bilayer membranes containing any sterol as well as sterol-free bilayer membranes due to the release of AmB and its incorporation within the membranes. However, a second phase of increasing fluorescence was found in the case of ergosterol alone. On the other hand, in the 47% monomeric form, AmB gave a biphasic intensity profile in membranes containing any sterol as well as sterol-free membranes. However, the extent of the second phase of increasing fluorescence intensity was markedly dependent upon sterol composition. Studies using sterol-containing vesicles provide important insights into the role of the aggregation state of AmB in its effects on cells.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anfotericina B, ~80% (HPLC), powder