Merck
  • Discovery of aminobenzyloxyarylamides as κ opioid receptor selective antagonists: application to preclinical development of a κ opioid receptor antagonist receptor occupancy tracer.
Discovery of aminobenzyloxyarylamides as κ opioid receptor selective antagonists: application to preclinical development of a κ opioid receptor antagonist receptor occupancy tracer.
Journal of medicinal chemistry (2011-10-01)
Charles H Mitch, Steven J Quimby, Nuria Diaz, Concepcion Pedregal, Marta G de la Torre, Alma Jimenez, Qing Shi, Emily J Canada, Steven D Kahl, Michael A Statnick, David L McKinzie, Dana R Benesh, Karen S Rash, Vanessa N Barth
RESUMO
Arylphenylpyrrolidinylmethylphenoxybenzamides were found to have high affinity and selectivity for κ opioid receptors. On the basis of receptor binding assays in Chinese hamster ovary (CHO) cells expressing cloned human opioid receptors, (S)-3-fluoro-4-(4-((2-(3-fluorophenyl)pyrrolidin-1-yl)methyl)phenoxy)benzamide (25) had a K(i) = 0.565 nM for κ opioid receptor binding while having a K(i) = 35.8 nM for μ opioid receptors and a K(i) = 211 nM for δ opioid receptor binding. Compound 25 was also a potent antagonist of κ opioid receptors when tested in vitro using a [(35)S]-guanosine 5'O-[3-thiotriphosphate] ([(35)S]GTP-γ-S) functional assay in CHO cells expressing cloned human opioid receptors. Compounds were also evaluated for potential use as receptor occupancy tracers. Tracer evaluation was done in vivo, using liquid chromatography-tandem mass spectrometry (LC/MS/MS) methods, precluding the need for radiolabeling. (S)-3-Chloro-4-(4-((2-(pyridine-3-yl)pyrrolidin-1-yl)methyl)phenoxy)benzamide (18) was found to have favorable properties for a tracer for receptor occupancy, including good specific versus nonspecific binding and good brain uptake.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
LY2444296, ≥98% (HPLC)