Merck
  • Regulation of tyrosine hydroxylase mRNA stability by protein-binding, pyrimidine-rich sequence in the 3'-untranslated region.
Regulation of tyrosine hydroxylase mRNA stability by protein-binding, pyrimidine-rich sequence in the 3'-untranslated region.
The Journal of biological chemistry (1999-01-16)
W R Paulding, M F Czyzyk-Krzeska
RESUMO
The stability of mRNA for tyrosine hydroxylase (TH), the rate-limiting enzyme in catecholamine synthesis, is regulated by oxygen tension in the pheochromocytoma-derived PC12 cell line. We previously identified a pyrimidine-rich 27-base-long protein-binding sequence in the 3'-untranslated region of TH mRNA that is associated with hypoxia-inducible formation of a ribonucleoprotein complex (hypoxia-inducible protein-binding site (HIPBS)). In this study, we show that HIPBS is an mRNA stabilizing element necessary for both constitutive and hypoxia-regulated stability of TH mRNA. The mutations within this sequence that abolish protein binding markedly decrease constitutive TH mRNA stability and ablate its hypoxic regulation. A short fragment of TH mRNA that contains the wild-type HIPBS confers the increased mRNA stability to the reporter chloramphenicol acetyltransferase mRNA. However, it is not sufficient to confer hypoxic regulation. The HIPBS element binds two isoforms of a 40-kDa poly(C)-binding protein (PCBP). Hypoxia induces expression of the isoform 1, PCBP1, but not the isoform 2, PCBP2, in PC12 cells.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Polycytidylic acid–Agarose, lyophilized powder