Merck
  • Tissue inhibitor of metalloproteinases-1 stimulates gene expression in MDA-MB-435 human breast cancer cells by means of its ability to inhibit metalloproteinases.
Tissue inhibitor of metalloproteinases-1 stimulates gene expression in MDA-MB-435 human breast cancer cells by means of its ability to inhibit metalloproteinases.
Breast cancer research and treatment (2005-09-06)
Joseph F Porter, Shashi Sharma, Donna L Wilson, Maya A Kappil, Ronald P Hart, David T Denhardt
RESUMO
Tissue inhibitor of metalloproteinases-1 (TIMP-1) is a widely expressed, secreted protein that functions primarily to inhibit members of a large family of metalloproteinases (MPs). Because of the ability of TIMP-1 to inhibit MPs, it functions in many of the same pathophysiological processes as these enzymes, e.g. wound healing, ovulation, angiogenesis, and cancer cell metastasis. TIMP-1 can also stimulate proliferation ([3H]thymidine incorporation) and cellular anabolic processes (Alamar Blue reduction). This stimulation has been shown to be dependent on the MP-inhibitory ability of TIMP-1 in the human breast cancer cell line MDA-MB-435 (Porter et al., Br J Cancer 90: 463, 2004). To shed light on the mechanism by which TIMP-1 stimulates cellular anabolic processes, an oligonucleotide microarray analysis was performed over a time course of TIMP-1 treatment of MDA-MB-435 cells. Fifteen genes whose mRNAs were differentially regulated were identified. Six (Importin-7, MGC10471, FOXC1, subunit p20 of Arp2/3 complex, mitochondrial ribosomal protein L32, and the serine/threonine kinase-4 (MST1)) of these genes were confirmed by quantitative real time PCR. These same mRNAs were shown to be regulated by the synthetic hydroxamate MP-inhibitor GM6001 but not by its inactive derivative GM6001*, suggesting that the differential regulation occurs through the MP-inhibitory ability of TIMP-1. These results suggest a complex action of TIMP-1 on cancer cells mediated by constitutively active cell surface metalloproteinases that release factors regulating cell signaling pathways; they may account for the paradoxical observation that elevated levels of TIMP-1 in tumors can correlate with an adverse prognosis.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
GM6001 MMP Inhibitor Powder, The GM6001 MMP Inhibitor Powder controls the biological activity of MMP. This small molecule/inhibitor is primarily used for Biochemicals applications.