Merck
  • The isolation and preliminary characterization of N-acetyl-D-glucosamine kinase from rat kidney and liver.
The isolation and preliminary characterization of N-acetyl-D-glucosamine kinase from rat kidney and liver.
The Biochemical journal (1980-03-01)
M B Allen, D G Walker
RESUMO
1. Procedures for the extensive purification in high yield of N-acetyl-D-glucosamine kinase from rat liver and kidney are described. The separation of this enzyme from hepatic glucokinase depended primarily on their differing behaviour on an affinity column of Sepharose--N-(6-aminohexanoyl)-2-amino-2-deoxy-D-glucopyranose. 2. This N-acetyl-D-glucosamine kinase also catalyses the phosphorylation of N-acetyl-D-mannosamine and, at a lower rate, several other sugar analogues, including D-glucose. 3. A comparison of the behaviour of the enzyme during gel filtration and electrophoresis in sodium dodecyl sulphate/polyacrylamide gels suggests that N-acetyl-D-glucosamine kinase is a symmetrical dimer of mol.wt. 80000.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
N-Acetyl-D-glucosamine–Agarose, saline suspension