Merck
Murine Frizzled-1 behaves as an antagonist of the canonical Wnt/beta-catenin signaling.
The Journal of biological chemistry (2003-11-25)
Sergio Roman-Roman, De-Li Shi, Véronique Stiot, Eric Haÿ, Béatrice Vayssière, Teresa Garcia, Roland Baron, Georges Rawadi
RESUMO
Activation of the Wnt signaling cascade provides key signals during development and in disease. Wnt signals are transduced by seven-transmembrane Frizzleds (Fzs) and the single transmembrane low density lipoprotein receptor-related proteins 5 or 6. In the course of the analysis of genes regulated by bone morphogenetic protein 2 in mesenchymal cells we found a significant induction of murine Frizzled-1 (mFz1) gene expression. Unexpectedly overexpression of mFz1 dramatically repressed the induction of alkaline phosphatase mediated by either bone morphogenetic protein 2 or Wnt3a in these cells. Moreover mFz1 overexpression significantly repressed both beta-catenin translocation into the nucleus and T cell factor signaling mediated by Wnt3a. Importantly microinjection of mFz1 transcript in Xenopus embryo inhibited the ability of Wnt1 to induce the expression of the Wnt/beta-catenin target gene Siamois in animal cap assay and secondary axis formation in whole embryo. By using chimeric constructs in which N- and C-terminal segments of mFz1 were replaced by the corresponding parts of Xfz3 we demonstrated that the antagonistic activity resides in the cysteine-rich domain of the N-terminal part. The antagonist activity of mFz1 could be prevented by overexpression of Galphaq-(305-359), which specifically uncouples Gq-coupled receptors, suggesting that Galphaq signaling contributes to the inhibition of Wnt/beta-catenin pathway by mFz1. This is the first time that a Frizzled receptor has been reported to antagonize Wnt/beta-catenin.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
TOPflash (TCF Reporter Plasmid), Transfection grade T-cell factor reporter plasmid containing 2 sets (with the second set in the reverse orientation) of 3 copies of the TCF binding site (wild type) upstream of the Thymidine Kinase minimal promoter and Luciferase open reading frame.