Merck
Downregulation of delta CaM kinase II in human tumor cells.
Biochimica et biophysica acta (1999-10-19)
R M Tombes, R B Mikkelsen, W D Jarvis, S Grant
RESUMO
Over two dozen alternative splice variants of CaMK-II, the type II Ca(2+)/CaM-dependent protein kinase, are encoded from four genes (alpha, beta, gamma and delta) in mammalian cells. Isozymes of alpha and beta CaMK-II are well characterized in brain; however, an understanding of the relative endogenous levels of CaMK-II isozymes in a wide variety of non-neuronal cells has not yet been described. In this study, we have demonstrated that CaMK-II consists primarily of the 54 kDa delta CaMK-II (delta(2) or delta(C)) isozyme in rodent fibroblasts. beta and gamma CaMK-II isozymes are minor and alpha CaMK-II was not expressed. The primary delta CaMK-II in human fibroblasts and the MCF10A mammary epithelial cell line was the 52 kDa delta(4) CaMK-II, an isozyme identical to delta(2) except for a missing 21-amino-acid C-terminal tail. delta CaMK-II levels were diminished in both human and rodent fibroblasts after SV40 transformation and in the mammary adenocarcinoma MCF7 cell line when compared to MCF10A cells. In fact, most tumor cells exhibited CaMK-II specific activities which were two- to tenfold lower than in untransformed fibroblasts. We conducted complementary CaMK-II studies on the NGF-induced differentiation of rat PC-12 cells. Although no new synthesis of CaMK-II occurs, neurite outgrowth in these cells is accompanied by a preferential activation of delta CaMK-II. Endogenous delta CaMK-II has a perinuclear distribution in fibroblasts and extends along neurites in PC-12 cells. These findings point to a role for delta CaMK-II isozymes in cellular differentiation.