Merck
Enhancing effect of albumin hydrolysate on ethanol production employing Saccharomyces sake.
Biotechnology and bioengineering (1995-03-05)
C S Shin, J Y Song, O H Ryu, S S Wang
RESUMO
The enhancing effect of albumin hydrolysate on ethanol production was investigated in ethanol fermentations using Saccharomyces sake. In batchwise ethanol production, addition of supplemental albumin hydrolysate and phosphatidylcholine, or albumin hydrolysate alone, brought about a more than 60% increase in final ethanol concentration (148 or 144 g/L compared with 88 g/L with no supplementation [control] after 72 h). The effect of the supplements is believed to be due to an enhanced alcohol tolerance of cells grown in media containing the supplements. Cells grown in media containing albumin hydrolysate were enriched in phenyalanine, tyrosine, and methionine in their plasma membranes. All three amino acids were also present in considerable amounts in the albumin hydrolysate. This fact suggests that the three amino acids, which are present in albumin hydrolysate, are incorporated into the plasma membranes of cells. Under ethanol production conditions in which only one amino acid among the components of albumin hydrolysate was excluded, namely phenlalanine, tyrosine, or methionine, significant reductions in ethanol production resulted.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Albumin hydrolysate