Merck
Simultaneous spectrophotometric determination of nitrite and nitrate by flow-injection analysis.
Talanta (1996-07-01)
M J Ahmed, C D Stalikas, S M Tzouwara-Karayanni, M I Karayannis
RESUMO
An automatic direct spectrophotometric method for the simultaneous determination of nitrite and nitrate by flow-injection analysis has been developed. Nitrite reacts with 3-nitroaniline in the presence of hydrochloric acid (0.96-1.8 M HCl or pH 0.5-0.7) to form a diazonium cation, which is subsequently coupled with N-(1-naphthyl)-ethylenediamine dihydrochloride to form a stable purple azo dye, the absorbance of which is measured at 535 nm. Nitrate is reduced on-line to nitrite in a copper-coated cadmium column which is then treated with azo dye reagent and the absorbance due to the sum of nitrite and nitrate is measured; nitrate is determined from the difference in absorbance values. A copper column incorporated into the reaction manifold before the copperised cadmium column not only improves the long-term accuracy, but also extends the life time of the copperised cadmium column. Various analytical parameters, such as effect of acidity (pH), flow rate, sample size, dispersion coefficient, time, temperature, reagent concentration and interfering species, were studied. The calibration graphs were rectilinear for 0.1-3.5 mug ml(-1) of NO(3) and 10 ng ml(-1)-2.2mug ml(-1) of NO(2). The method is successfully applied to some food samples (meat, flour and cheese), environmental waters (inland and surface), beer and soil samples. Up to 30 samples can be analysed per hour with a relative precision of approximately 0.1-2%.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
N-(1-Naphthyl)ethylenediamine dihydrochloride monomethanolate, for spectrophotometric det. of nitrate and nitrite, ≥99.0%
Preço e disponibilidade não estão disponíveis no momento.