Merck
Tumor-targeting prodrug-activating bacteria for cancer therapy.
Cancer gene therapy (2008-03-29)
C-M Cheng, Y-L Lu, K-H Chuang, W-C Hung, J Shiea, Y-C Su, C-H Kao, B-M Chen, S Roffler, T-L Cheng
RESUMO
Increasing the specificity of chemotherapy may improve the efficacy of cancer treatment. Toward this aim, we developed a strain of bacteria to express enzymes for selective prodrug activation and non-invasive imaging in tumors. beta-glucuronidase and the luxCDABE gene cluster were expressed in the DH5alpha strain of Escherichia coli to generate DH5alpha-lux/betaG. These bacteria emitted light for imaging and hydrolyzed the glucuronide prodrug 9ACG to the topoisomerase I inhibitor 9-aminocamptothecin (9AC). By optical imaging, colony-forming units (CFUs) and staining for betaG activity, we found that DH5alpha-lux/betaG preferentially localized and replicated within CL1-5 human lung tumors in mice. The intensity of luminescence, CFU and betaG activity increased with time, indicating bacterial replication occurred in tumors. In comparison with DH5alpha-lux/betaG, 9AC or 9ACG treatment, combined systemic administration of DH5alpha-lux/betaG followed by 9ACG prodrug treatment significantly (P<0.005) delayed the growth of CL1-5 tumors. Our results demonstrate that prodrug-activating bacteria may be useful for selective cancer chemotherapy.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
β-Glucuronidase Reporter Gene Staining Kit