Merck
  • Discovery of a series of imidazo[4,5-b]pyridines with dual activity at angiotensin II type 1 receptor and peroxisome proliferator-activated receptor-γ.
Discovery of a series of imidazo[4,5-b]pyridines with dual activity at angiotensin II type 1 receptor and peroxisome proliferator-activated receptor-γ.
Journal of medicinal chemistry (2011-05-12)
Agustin Casimiro-Garcia, Gary F Filzen, Declan Flynn, Christopher F Bigge, Jing Chen, Jo Ann Davis, Danette A Dudley, Jeremy J Edmunds, Nadia Esmaeil, Andrew Geyer, Ronald J Heemstra, Mehran Jalaie, Jeffrey F Ohren, Robert Ostroski, Teresa Ellis, Robert P Schaum, Chad Stoner
RESUMO
Mining of an in-house collection of angiotensin II type 1 receptor antagonists to identify compounds with activity at the peroxisome proliferator-activated receptor-γ (PPARγ) revealed a new series of imidazo[4,5-b]pyridines 2 possessing activity at these two receptors. Early availability of the crystal structure of the lead compound 2a bound to the ligand binding domain of human PPARγ confirmed the mode of interaction of this scaffold to the nuclear receptor and assisted in the optimization of PPARγ activity. Among the new compounds, (S)-3-(5-(2-(1H-tetrazol-5-yl)phenyl)-2,3-dihydro-1H-inden-1-yl)-2-ethyl-5-isobutyl-7-methyl-3H-imidazo[4,5-b]pyridine (2l) was identified as a potent angiotensin II type I receptor blocker (IC(50) = 1.6 nM) with partial PPARγ agonism (EC(50) = 212 nM, 31% max) and oral bioavailability in rat. The dual pharmacology of 2l was demonstrated in animal models of hypertension (SHR) and insulin resistance (ZDF rat). In the SHR, 2l was highly efficacious in lowering blood pressure, while robust lowering of glucose and triglycerides was observed in the male ZDF rat.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
5-Bromo-1-indanone, 97%