Merck
SUMO, ubiquitin's mysterious cousin.
Nature reviews. Molecular cell biology (2001-03-27)
S Müller, C Hoege, G Pyrowolakis, S Jentsch
RESUMO
Covalent modification of cellular proteins by the ubiquitin-like modifier SUMO regulates various cellular processes, such as nuclear transport, signal transduction, stress response and cell-cycle progression. But, in contrast to ubiquitylation, sumoylation does not tag proteins for degradation, but seems to enhance their stability or modulate their subcellular compartmentalization.