Merck
  • Conformational and molecular basis for induction of apoptosis by a p53 C-terminal peptide in human cancer cells.
Conformational and molecular basis for induction of apoptosis by a p53 C-terminal peptide in human cancer cells.
The Journal of biological chemistry (1999-11-27)
A L Kim, A J Raffo, P W Brandt-Rauf, M R Pincus, R Monaco, P Abarzua, R L Fine
RESUMO
A p53-derived C-terminal peptide induced rapid apoptosis in breast cancer cell lines carrying endogenous p53 mutations or overexpressed wild-type (wt) p53 but was not toxic to nonmalignant human cell lines containing wt p53. Apoptosis occurred through a Fas/APO-1 signaling pathway involving increased extracellular levels of Fas/FasL in the absence of protein synthesis, as well as activation of a Fas/APO-1-specific protease, FLICE. The peptide activity was p53-dependent, and it had no effect in three tumor cell lines with null p53. Furthermore, the C-terminal peptide bound to p53 protein in cell extracts. Thus, p53-dependent, Fas/APO-1 mediated apoptosis can be induced in breast cancer cells with mutant p53 similar to the recently described Fas/APO-1 induced apoptosis by wt p53. However, mutant p53 without p53 peptide does not induce a Fas/APO-1 activation or apoptosis. Docking of the computed low energy conformations for the C-terminal peptide with those for a recently defined proline-rich regulatory region from the N-terminal domain of p53 suggests a unique low energy complex between the two peptide domains. The selective and rapid induction of apoptosis in cancer cells carrying p53 abnormalities may lead to a novel therapeutic modality.