Merck
  • Human bile reduces antimicrobial activity of selected antibiotics against Enterococcus faecalis and Escherichia coli in vitro
Human bile reduces antimicrobial activity of selected antibiotics against Enterococcus faecalis and Escherichia coli in vitro
Antimicrobial agents and chemotherapy (2017)
Wulkersdorfer B, et al.