Merck
  • AR-13 reduces antibiotic-resistant bacterial burden in cystic fibrosis phagocytes and improves cystic fibrosis transmembrane conductance regulator function
AR-13 reduces antibiotic-resistant bacterial burden in cystic fibrosis phagocytes and improves cystic fibrosis transmembrane conductance regulator function
Journal of Cystic Fibrosis : Official Journal of the European Cystic Fibrosis Society (2018)
Assani K, et al.