Merck
  • Blood Telomere Length Changes After Ritonavir-Boosted Darunavir Combined With Raltegravir or Tenofovir-Emtricitabine in Antiretroviral-Naive Adults Infected With HIV-1.
Blood Telomere Length Changes After Ritonavir-Boosted Darunavir Combined With Raltegravir or Tenofovir-Emtricitabine in Antiretroviral-Naive Adults Infected With HIV-1.
The Journal of infectious diseases (2018-07-10)
Natalia Stella-Ascariz, Rocio Montejano, Javier Rodriguez-Centeno, Belen Alejos, Christine Schwimmer, Jose I Bernardino, Berta Rodes, Clotilde Allavena, Christian Hoffmann, Magnus Gisslén, Rosa de Miguel, Andrés Esteban-Cantos, Cédrick Wallet, François Raffi, Jose R Arribas
RESUMO
Tenofovir is a potent inhibitor of human telomerase. The clinical relevance of this inhibition is unknown. NEAT001/ANRS143 is a randomized trial that showed noninferiority over 96 weeks of ritonavir-boosted darunavir plus raltegravir versus tenofovir disoproxil fumarate/emtricitabine in 805 antiretroviral antiretrovrial-naive HIV-infected adults. We compared changes in whole-blood telomere length measured with quantitative polymerase chain reaction in 201 randomly selected participants (104 raltegravir and 97 tenofovir disoproxil fumarate/emtricitabine). We performed multivariable estimative and predictive linear regression. At week 96, participants receiving tenofovir disoproxil fumarate/emtricitabine had a statistically significant higher gain in telomere length than participants receiving raltegravir. Difference in mean telomere length change between groups (tenofovir disoproxil fumarate/emtricitabine minus raltegravir) from baseline to week 96 adjusted by baseline telomere length was 0.031 (P = .009). This difference was not significantly confounded by age, gender, known duration of HIV infection, CD4 (baseline/nadir), CD8 cells, CD4/CD8 ratio, HIV viral load (baseline/week 96), tobacco and alcohol consumption, statins, or hepatitis C. Antiretroviral-naive HIV-infected adults receiving ritonavir-boosted darunavir and tenofovir disoproxil fumarate/emtricitabine had a significant higher gain in blood telomere length than those receiving ritonavir-boosted darunavir and raltegravir, suggesting a better initial recovery from HIV-associated immunosenescence.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Darunavir, ≥98% (HPLC)