Merck
  • Toward the Synthesis of Nuphar Sesquiterpene Thioalkaloids: Stereodivergent Rhodium-Catalyzed Synthesis of the Thiolane Subunit.
Toward the Synthesis of Nuphar Sesquiterpene Thioalkaloids: Stereodivergent Rhodium-Catalyzed Synthesis of the Thiolane Subunit.
The Journal of organic chemistry (2015-07-07)
Ping Lu, Aaron T Herrmann, Armen Zakarian
RESUMO
A stereodivergent approach to the central thiolane subunit of Nuphar sesquiterpene thioalkaloids has been developed. This approach features a rhodium-catalyzed Stevens-type rearrangement in conjunction with an enzyme resolution reaction. Further elaboration into a polycyclic ring system via alcohol oxidation and ring-closing metathesis is also described.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Rh2(R-PTAD)4
Sigma-Aldrich
Rh2(S-PTAD)4