Merck
  • Repetitive acute intermittent hypoxia increases growth/neurotrophic factor expression in non-respiratory motor neurons.
Repetitive acute intermittent hypoxia increases growth/neurotrophic factor expression in non-respiratory motor neurons.
Neuroscience (2016-03-06)
I Satriotomo, N L Nichols, E A Dale, A T Emery, J M Dahlberg, G S Mitchell
RESUMO
Repetitive acute intermittent hypoxia (rAIH) increases growth/trophic factor expression in respiratory motor neurons, thereby eliciting spinal respiratory motor plasticity and/or neuroprotection. Here we demonstrate that rAIH effects are not unique to respiratory motor neurons, but are also expressed in non-respiratory, spinal alpha motor neurons and upper motor neurons of the motor cortex. In specific, we used immunohistochemistry and immunofluorescence to assess growth/trophic factor protein expression in spinal sections from rats exposed to AIH three times per week for 10weeks (3×wAIH). 3×wAIH increased brain-derived neurotrophic factor (BDNF), its high-affinity receptor, tropomyosin receptor kinase B (TrkB), and phosphorylated TrkB (pTrkB) immunoreactivity in putative alpha motor neurons of spinal cervical 7 (C7) and lumbar 3 (L3) segments, as well as in upper motor neurons of the primary motor cortex (M1). 3×wAIH also increased immunoreactivity of vascular endothelial growth factor A (VEGFA), the high-affinity VEGFA receptor (VEGFR-2) and an important VEGF gene regulator, hypoxia-inducible factor-1α (HIF-1α). Thus, rAIH effects on growth/trophic factors are characteristic of non-respiratory as well as respiratory motor neurons. rAIH may be a useful tool in the treatment of disorders causing paralysis, such as spinal injury and motor neuron disease, as a pretreatment to enhance motor neuron survival during disease, or as preconditioning for cell-transplant therapies.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 (KDR) antibody produced in mouse, clone 260.4, KDR-2, ascites fluid