Merck
  • Clinical Utility of Classical and Nonclassical Monocyte Percentage in the Diagnosis of Chronic Myelomonocytic Leukemia.
Clinical Utility of Classical and Nonclassical Monocyte Percentage in the Diagnosis of Chronic Myelomonocytic Leukemia.
American journal of clinical pathology (2018-07-10)
Chad A Hudson, W Richard Burack, Patricia C Leary, John M Bennett
RESUMO
To determine if a clinically applicable flow cytometry methodology could identify chronic myelomonocytic leukemia (CMML) cases. Monocyte subset screening (CD14/CD16 expression) was performed on 68 blood and 25 bone marrow specimens with a monocytosis and/or flagged as possible CMML. Fifty thousand total events were obtained per case. Cases were categorized as CMML, atypical chronic myeloid leukemia (aCML), or non-CMML + non-aCML by clinicopathologic diagnosis. The methodology differentiated blood and bone marrow CMML cases from non-CMML + non-aCML but not three aCML cases in the clinical setting. Furthermore, a decreased percentage of nonclassical monocytes (CD14dimCD16+) showed better sensitivity than the previously described approach that relied on increased percentage of classical monocytes (CD14brightCD16-). Quantification of monocyte subsets is useful in clinical practice as a diagnostic marker of CMML in blood and bone marrow specimens. The percentage of nonclassical monocytes should be included in analysis of monocyte subsets.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Mordant Orange 1, Dye content 70 %