Merck
  • The EZH2- H3K27me3-DNMT1 complex orchestrates epigenetic silencing of the wwc1 gene, a Hippo/YAP pathway upstream effector, in breast cancer epithelial cells.
The EZH2- H3K27me3-DNMT1 complex orchestrates epigenetic silencing of the wwc1 gene, a Hippo/YAP pathway upstream effector, in breast cancer epithelial cells.
Cellular signalling (2018-08-20)
Xiong Liu, Chun Li, Rendong Zhang, Wenjun Xiao, Xia Niu, Xiajun Ye, Zijia Li, Yuxian Guo, Junyu Tan, Yaochen Li
RESUMO
It is well known that epithelial-mesenchymal transition (EMT) can confer cancer cells with invasive and migratory capabilities associated with distant metastasis. As a key upstream factor in the Hippo pathway, Kibra (wwc1 gene) has been shown to suppress EMT in breast cancer cells, and we have found that its expression is reduced or lost completely in both human breast cancer cell lines and clinical tissue samples, particularly in triple negative breast cancer (TNBC). Unfortunately, the molecular mechanisms underlying this progression-associated event remain to be elucidated. Epigenetic gene silencing is one of the most common causes of suppressed expression of tumor suppressor genes. Furthermore, recent studies have demonstrated that EZH2 can recruit DNA methyltransferases, resulting in DNA methylation and subsequent gene silencing in certain circumstances. Thus, we hypothesized that there may exist a link between EZH2 and DNA methylation in association with wwc1 silencing in breast cancer. To test this hypothesis, we performed bisulfite sequencing, shRNA, co-IP, ChIP, MeDIP and ChIP-qPCR. As expected, RG108 or 5-Aza treatment improved the wwc1 gene transcription and Kibra protein expression. Both bisulfite sequencing and MeDIP demonstrated higher CpG methylation of the wwc1 promoter the TNBC cells (MDA-MB-231) than in luminal breast cancer cells (MCF7). It is noteworthy that ChIP and co-IP assays showed that EZH2, H3K27me3 and DNMT1 are enriched at the wwc1 promoter, and there exist physiologically relevant protein-protein interactions between them. We also found that EZH2 knockdown leads to a partial increase in Kibra expression and a considerable reduction in H3K27 and DNMT1 trimethylation. Moreover, ChIP-qPCR revealed more DNA fragments containing the wwc1 promoter in MDA-MB-231 than in MCF7 cells after immunoprecipitation with EZH2, DNMT1 and H3K27me3 antibodies. Collectively, our results reveal crosstalk between H3K27me3 inhibition catalyzed by EZH2 and CpG island methylation mediated by DNMT1 within the wwc1 promoter, which synergistically silence wwc1 gene expression in TNBC. Based on these results, we conclude that EZH2 shows promise as a potential anti-tumor target.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
RG108, ≥98% (HPLC), powder