Merck
Genetic variants in the VEGF pathway as prognostic factors in stages II and III colon cancer.
The pharmacogenomics journal (2017-12-29)
Pau Riera, Anna C Virgili, Juliana Salazar, Ana Sebio, María Tobeña, Ivana Sullivan, David Páez
RESUMO
The role of vascular endothelial growth factor (VEGF) gene polymorphisms in the prognosis of colon cancer prognosis remains unclear. We evaluated the influence of 28 single-nucleotide polymorphisms in 12 genes in the VEGF pathway on the prognosis of 347 patients with stage II-III colon cancer. We found that rs9513070 (VEGFR1) and rs1137282 (KRAS) were associated with overall survival in stage II colon cancer patients (p = 0.025 and p = 0.001, respectively). When primary tumor location was considered, rs9513070 was also associated with relapse-free and overall survival (p = 0.033 and p = 0.031, respectively) in left colon cancer patients. Additionally, rs35251833 in the ITGAV gene correlated with relapse-free survival (p = 0.032). This study provides evidence that germline polymorphisms in VEGFR1, KRAS and ITGAV genes are associated with prognosis in stages II-III colon cancer patients. As stage and tumor location are correlated with prognosis, future genetic studies should stratify colon cancer patients according to these parameters.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Metastin (45-54) amide, human, ≥95%