Merck
  • Radical Trifluoromethoxylation of Arenes Triggered by a Visible-Light-Mediated N-O Bond Redox Fragmentation.
Radical Trifluoromethoxylation of Arenes Triggered by a Visible-Light-Mediated N-O Bond Redox Fragmentation.
Angewandte Chemie (International ed. in English) (2018-06-22)
Benson J Jelier, Pascal F Tripet, Ewa Pietrasiak, Ivan Franzoni, Gunnar Jeschke, Antonio Togni
RESUMO
A simple trifluoromethoxylation method enables non-directed functionalization of C-H bonds on a range of substrates, providing access to aryl trifluoromethyl ethers. This light-driven process is distinctly different from conventional procedures and occurs through an OCF3 radical mechanism mediated by a photoredox catalyst, which triggers an N-O bond fragmentation. The pyridinium-based trifluoromethoxylation reagent is bench-stable and provides access to synthetic diversity in lead compounds in an operationally simple manner.