Merck
  • Effective methodologies for enantiomeric separations of 150 pharmacology and toxicology related 1°, 2°, and 3° amines with core-shell chiral stationary phases.
Effective methodologies for enantiomeric separations of 150 pharmacology and toxicology related 1°, 2°, and 3° amines with core-shell chiral stationary phases.
Journal of pharmaceutical and biomedical analysis (2018-04-07)
Garrett Hellinghausen, Daipayan Roy, Jauh T Lee, Yadi Wang, Choyce A Weatherly, Diego A Lopez, Kate A Nguyen, John D Armstrong, Daniel W Armstrong
RESUMO
Core-shell particles (superficially porous particles, SPPs) have been proven to provide high-throughput and effective separations of a variety of chiral molecules. However, due to their limited commercialization, many separations have not been reported with these stationary phases. In this study, four SPP chiral stationary phases (CSPs) were utilized for the enantiomeric separation of 150 chiral amines. These amines encompass a variety of structural and drug classes, which are particularly important to the pharmaceutical industry and in forensics. This comprehensive evaluation demonstrates the power of these CSPs and the ease of method development and optimization. The CSPs used in this study included the macrocyclic glycopeptide-based CSPs (VancoShell and NicoShell), the cyclodextrin-based CSP (CDShell-RSP), and the cyclofructan-based CSP (LarihcShell-P). These CSPs offered versatility for a variety of applications and worked in a complementary fashion to baseline separate all 150 amines. The LarihcShell-P was highly effective for separating primary amines. VancoShell, NicoShell, and CDShell-RSP were useful for separating all types of amines. These CSPs are multi-modal and can be utilized with mass spectrometry compatible solvents. Eighteen racemic controlled substances were simultaneously baseline separated in a single liquid chromatography-mass spectrometry (LC-MS) analysis. Details in high-performance liquid chromatography (HPLC) parameters will be discussed as well as the improved chromatographic performance afforded by the SPP CSPs.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Ammonium trifluoroacetate, 98%