Merck
  • Hepatitis C virus core protein triggers abnormal porphyrin metabolism in human hepatocellular carcinoma cells.
Hepatitis C virus core protein triggers abnormal porphyrin metabolism in human hepatocellular carcinoma cells.
PloS one (2018-06-02)
Takafumi Nakano, Kyoji Moriya, Kazuhiko Koike, Toshiharu Horie
RESUMO
Porphyria cutanea tarda (PCT), the most common of the human porphyrias, arises from a deficiency of uroporphyrinogen decarboxylase. Studies have shown a high prevalence of hepatitis C virus (HCV) infection in patients with PCT. While these observations implicate HCV infection as a risk factor for PCT pathogenesis, the mechanism of interaction between the virus and porphyrin metabolism is unknown. This study aimed to assess the effect of HCV core protein on intracellular porphyrin metabolism to elucidate the link between HCV infection and PCT. The accumulation and excretion of porphyrins after treatment with 5-aminolevulinic acid, a porphyrin precursor, were compared between cells stably expressing HCV core protein and controls. Cells expressing HCV core protein had lower amounts of intracellular protoporphyrin IX and heme and had higher amounts of excreted coproporphyrin III, the oxidized form of coproporphyrinogen III, compared with controls. These observations suggest that HCV core protein affects porphyrin metabolism and facilitates the export of excess coproporphyrinogen III and/or coproporphyrin III, possibly via porphyrin transporters. Real-time PCR analysis revealed that the presence of HCV core protein increased the mRNA expression of porphyrin exporters ABCG2 and FLVCR1. Western blot analysis showed a higher expression level of FLVCR1, but not ABCG2, as well as a higher expression level of mature ALAS1, which is the rate-limiting enzyme in the heme synthesis pathway, in HCV core protein-expressing cells compared with controls. The data indicate that HCV core protein induced abnormal intracellular porphyrin metabolism, with an over-excretion of coproporphyrin III. These findings may partially account for the susceptibility of HCV-infected individuals to PCT development.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Protoporphyrin IX, ≥95%