Merck
Pb(II) Induces Scramblase Activation and Ceramide-Domain Generation in Red Blood Cells.
Scientific reports (2018-05-12)
Hasna Ahyayauch, Aritz B García-Arribas, Jesús Sot, Emilio J González-Ramírez, Jon V Busto, Bingen G Monasterio, Noemi Jiménez-Rojo, F Xabier Contreras, Adela Rendón-Ramírez, Cesar Martin, Alicia Alonso, Félix M Goñi
RESUMO
The mechanisms of Pb(II) toxicity have been studied in human red blood cells using confocal microscopy, immunolabeling, fluorescence-activated cell sorting and atomic force microscopy. The process follows a sequence of events, starting with calcium entry, followed by potassium release, morphological change, generation of ceramide, lipid flip-flop and finally cell lysis. Clotrimazole blocks potassium channels and the whole process is inhibited. Immunolabeling reveals the generation of ceramide-enriched domains linked to a cell morphological change, while the use of a neutral sphingomyelinase inhibitor greatly delays the process after the morphological change, and lipid flip-flop is significantly reduced. These facts point to three major checkpoints in the process: first the upstream exchange of calcium and potassium, then ceramide domain formation, and finally the downstream scramblase activation necessary for cell lysis. In addition, partial non-cytotoxic cholesterol depletion of red blood cells accelerates the process as the morphological change occurs faster. Cholesterol could have a role in modulating the properties of the ceramide-enriched domains. This work is relevant in the context of cell death, heavy metal toxicity and sphingolipid signaling.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
GW4869, ≥90% (NMR)