Merck
  • Pappalysin-1 T cell receptor transgenic allo-restricted T cells kill Ewing sarcoma in vitro and in vivo.
Pappalysin-1 T cell receptor transgenic allo-restricted T cells kill Ewing sarcoma in vitro and in vivo.
Oncoimmunology (2017-03-28)
Andreas Kirschner, Melanie Thiede, Thomas G P Grünewald, Rebeca Alba Rubio, Günther H S Richter, Thomas Kirchner, Dirk H Busch, Stefan Burdach, Uwe Thiel
RESUMO
Pregnancy-associated plasma protein-A (PAPPA), also known as pappalysin, is a member of the insulin-like growth factor (IGF) family. PAPPA acts as a protease, cleaving IGF inhibitors, i.e., IGF binding proteins (IGFBPs), thereby setting free IGFs. The insulin/IGF-axis is involved in cancer in general and in Ewing sarcoma (ES) in particular. ES is a highly malignant bone tumor characterized by early metastatic spread. PAPPA is associated with various cancers. It is overexpressed and required for proliferation in ES. PAPPA also stimulates normal bone growth. We isolated HLA-A*02:01+/peptide-restricted T cells from A*02:01- healthy donors directed against PAPPA, generated by priming with A*02:01+ PAPPA peptide loaded dendritic cells. After TCR identification, retrovirally TCR transduced CD8+ T cells were assessed for their in vitro specificity and in vivo efficacy in human ES bearing Rag2-/-γc-/- mice. Engraftment in mice and tumor infiltration of TCR transgenic T cells in the mice was evaluated. The TCR transgenic T cell clone PAPPA-2G6 demonstrated specific reactivity toward HLA-A*02:01+/PAPPA+ ES cell lines. We furthermore detected circulating TCR transgenic T cells in the blood in Rag2-/-γc-/- mice and in vivo engraftment in bone marrow. Tumor growth in mice with xenografted ES was significantly reduced after treatment with PAPPA-2G6 TCR transgenic T cells in contrast to controls. Tumors of treated mice revealed tumor-infiltrating PAPPA-2G6 TCR transgenic T cells. In summary, we demonstrate that PAPPA is a first-rate target for TCR-based immunotherapy of ES.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-PAPPA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution