Merck
  • Mechanisms underlying the sparing of masticatory versus limb muscle function in an experimental critical illness model.
Mechanisms underlying the sparing of masticatory versus limb muscle function in an experimental critical illness model.
Physiological genomics (2011-10-20)
Sudhakar Aare, Julien Ochala, Holly S Norman, Peter Radell, Lars I Eriksson, Hanna Göransson, Yi-Wen Chen, Eric P Hoffman, Lars Larsson
RESUMO
Acute quadriplegic myopathy (AQM) is a common debilitating acquired disorder in critically ill intensive care unit (ICU) patients that is characterized by tetraplegia/generalized weakness of limb and trunk muscles. Masticatory muscles, on the other hand, are typically spared or less affected, yet the mechanisms underlying this striking muscle-specific difference remain unknown. This study aims to evaluate physiological parameters and the gene expression profiles of masticatory and limb muscles exposed to factors suggested to trigger AQM, such as mechanical ventilation, immobilization, neuromuscular blocking agents, corticosteroids (CS), and sepsis for 5 days by using a unique porcine model mimicking the ICU conditions. Single muscle fiber cross-sectional area and force-generating capacity, i.e., maximum force normalized to fiber cross-sectional area (specific force), revealed maintained masseter single muscle fiber cross-sectional area and specific-force after 5 days' exposure to all triggering factors. This is in sharp contrast to observations in limb and trunk muscles, showing a dramatic decline in specific force in response to 5 days' exposure to the triggering factors. Significant differences in gene expression were observed between craniofacial and limb muscles, indicating a highly complex and muscle-specific response involving transcription and growth factors, heat shock proteins, matrix metalloproteinase inhibitor, oxidative stress responsive elements, and sarcomeric proteins underlying the relative sparing of cranial vs. spinal nerve innervated muscles during exposure to the ICU intervention.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-p62/SQSTM1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution