Merck
Percutaneous penetration modifiers and formulation effects.
International journal of pharmaceutics (2009-11-11)
Diksha Kaushik, Aurora Costache, Bozena Michniak-Kohn
RESUMO
The enhancement/retardation of percutaneous permeation of diethyl-m-toluamide (DEET) in the presence of five percutaneous penetration modifiers (laurocapram, 3-dodecanoyloxazolidin-2-one (N-0915), S,S-dimethyl-N-(4-bromobenzoyl) iminosulfurane (DMBIS), S,S-dimethyl-N-(2-methoxycarbonylbenzenesulfonyl) iminosulfurane (DMMCBI) and tert-butyl 1-dodecyl-2-oxoazepan-3-yl-carbamate (TBDOC)) was investigated. These permeation modifiers were formulated in either water, propylene glycol (PG), ethanol or polyethylene glycol 400 (PEG 400). The permeation studies indicated that laurocapram enhanced DEET permeation in PG, but retarded in PEG 400. Likewise, N-0915 acted as a retardant with ethanol and PEG 400, but not with water. DMBIS decreased the permeation with ethanol as compared to permeation with water, PEG 400 or PG. Similarly, DMMCB acted as a retardant with ethanol and PEG 400, but not with water or PG. TBDOC formulations revealed its activity as a retardant with ethanol, but behaved as enhancer with water, PG and PEG 400. In addition, penetration modifier interactions with stratum corneum ceramide were investigated using chemical modeling. This investigation is significant since it confirms the role of pharmaceutical formulations and shows for the first time that an enhancer can become a retardant or vice versa depending upon the vehicle in which it is applied to the skin. Hence, we should be using the term "penetration modifiers" for all such compounds.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
NSC43067, ≥98% (HPLC)