Voltar
Kern PNJ series precision balances
Products
Categoria de produto
balances (6)
Fabricante
Diâmetro (mm)
Capacidade de pesagem (g)
Tamanho do pacote
Filtrar produtos
Showing 1-6 of 6
Comparar
Número do produto
Descrição
Preços
Z742875
Kern PNJ series precision balances,
Kern PNJ 600-3M, weighing capacity 620 g, accuracy: 0.001 g, precision: ±0.004 g
Expandir
Ver preços
Z742878
Kern PNJ series precision balances,
Kern PNJ 3000-2M verified 965-216, weighing capacity 3200 g, accuracy: 0.01 g, precision: ±0.02 g, (Verification certificate included)
Expandir
Ver preços
Z742877
Kern PNJ series precision balances,
Kern PNJ 3000-2M, weighing capacity 3200 g, accuracy: 0.01 g, precision: ±0.02 g
Expandir
Ver preços
Z742876
Kern PNJ series precision balances,
Kern PNJ 600-3M verified 965-201, weighing capacity 620 g, accuracy: 0.001 g, precision: ±0.004 g g, (Verification certificate included)
Expandir
Ver preços
Z742881
Kern PNJ series precision balances,
Kern PNJ 12000-1M verified 965-217, weighing capacity 12000 g, accuracy: 0.1 g, precision: ±0.2 g, (Verification certificate included)
Expandir
Ver preços
Z742879
Kern PNJ series precision balances,
Kern PNJ 12000-1M, weighing capacity 12000 g, accuracy: 0.1 g, precision: ±0.2 g