Voltar
Kern EW-N series toploader balances
Products
Categoria de produto
balances (8)
Fabricante
Diâmetro (mm)
Capacidade de pesagem (g)
Resolução (g)
Tamanho do pacote
Filtrar produtos
Showing 1-8 of 8
Comparar
Número do produto
Descrição
Preços
Z742779
Kern EW-N series toploader balances,
Kern EW 2200-2NM+YKA-03, weighing capacity 2200 g, resolution: ±0.01 g, linearity: 0.01 g
Expandir
Ver preços
Z742784
Kern EW-N series toploader balances,
Kern EW 6200-2NM+YKA-03, weighing capacity 6200 g, resolution: ±0.03 g, linearity: 0.01 g
Expandir
Ver preços
Z742774
Kern EW-N series toploader balances,
Kern EW 620-3NM+YKA-03, weighing capacity 620 g, resolution: ±0.003 g, linearity: 0.001 g
Expandir
Ver preços
Z742786
Kern EW-N series toploader balances,
Kern EW 12000-1NM+YKA-03, weighing capacity 12000 g, resolution: ±0.2 g, linearity: 0.1 g
Expandir
Ver preços
Z742770
Kern EW-N series toploader balances,
Kern EW 220-3NM+YKA-03, weighing capacity 220 g, resolution: ±0.002 g, linearity: 0.002 g
Expandir
Ver preços
Z742777
Kern EW-N series toploader balances,
Kern EW 820-2NM+YKA-03, weighing capacity 820 g, resolution: ±0.01 g, linearity: 0.01 g
Expandir
Ver preços
Z742772
Kern EW-N series toploader balances,
Kern EW 420-3NM+YKA-03, weighing capacity 420 g, resolution: ±0.003 g, linearity: 0.001 g
Expandir
Ver preços
Z742781
Kern EW-N series toploader balances,
Kern EW 4200-2NM+YKA-03, weighing capacity 4200 g, resolution: ±0.02 g, linearity: 0.01 g