Skip to Content
Merck
All Photos(1)

Key Documents

V900712

Sigma-Aldrich

DL-Proline

Vetec, reagent grade, 98%

Synonym(s):

(±)-Pyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
80812
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

208 °C (dec.) (lit.)

SMILES string

OC(=O)C1CCCN1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)

InChI key

ONIBWKKTOPOVIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antoinette Bugyei-Twum et al.
Cardiovascular diabetology, 13, 89-89 (2014-06-03)
Despite advances in the treatment of heart failure, mortality remains high, particularly in individuals with diabetes. Activated transforming growth factor beta (TGF-β) contributes to the pathogenesis of the fibrotic interstitium observed in diabetic cardiomyopathy. We hypothesized that high glucose enhances
Souhir Marsit et al.
Molecular biology and evolution, 32(7), 1695-1707 (2015-03-10)
Although an increasing number of horizontal gene transfers have been reported in eukaryotes, experimental evidence for their adaptive value is lacking. Here, we report the recent transfer of a 158-kb genomic region between Torulaspora microellipsoides and Saccharomyces cerevisiae wine yeasts
Nina Maeshima et al.
The Journal of biological chemistry, 290(21), 13440-13453 (2015-04-04)
Lipid A in LPS activates innate immunity through the Toll-like receptor 4 (TLR4)-MD-2 complex on host cells. Variation in lipid A has significant consequences for TLR4 activation and thus may be a means by which Gram-negative bacteria modulate host immunity.
Ying-Nan Wu et al.
Nanomedicine : nanotechnology, biology, and medicine, 10(7), 1507-1516 (2014-04-29)
To reproduce a complex and functional tissue, it is crucial to provide a biomimetic cellular microenvironment that not only incorporates biochemical cues, but also physical features including the nano-topographical patterning, for cell/matrix interaction. We developed spatially-controlled nano-topography in the form
Jingjing Zhang et al.
Biomedical materials (Bristol, England), 9(3), 035011-035011 (2014-05-14)
Different forms of biomaterials, including microspheres, sponges, hydrogels and nanofibres have been broadly used in cartilage regeneration; however, effects of internal structures of biomaterials on chondrogenesis of mesenchymal stem cells (MSCs) remain largely unexplored. Here we investigated the effect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service