Merck
  • Substrate topography determines the fate of chondrogenesis from human mesenchymal stem cells resulting in specific cartilage phenotype formation.
Substrate topography determines the fate of chondrogenesis from human mesenchymal stem cells resulting in specific cartilage phenotype formation.
Nanomedicine : nanotechnology, biology, and medicine (2014-04-29)
Ying-Nan Wu, Jaslyn Bee Khuan Law, Ai Yu He, Hong Yee Low, James H P Hui, Chwee Teck Lim, Zheng Yang, Eng Hin Lee
ABSTRACT
To reproduce a complex and functional tissue, it is crucial to provide a biomimetic cellular microenvironment that not only incorporates biochemical cues, but also physical features including the nano-topographical patterning, for cell/matrix interaction. We developed spatially-controlled nano-topography in the form of nano-pillar, nano-hole and nano-grill on polycaprolactone surface via thermal nanoimprinting. The effects of chondroitin sulfate-coated nano-topographies on cell characteristics and chondrogenic differentiation of human mesenchymal stem cell (MSC) were investigated. Our results show that various nano-topographical patterns triggered changes in MSC morphology and cytoskeletal structure, affecting cell aggregation and differentiation. Compared to non-patterned surface, nano-pillar and nano-hole topography enhanced MSC chondrogenesis and facilitated hyaline cartilage formation. MSCs experienced delayed chondrogenesis on nano-grill topography and were induced to fibro/superficial zone cartilage formation. This study demonstrates the sensitivity of MSC differentiation to surface nano-topography and highlights the importance of incorporating topographical design in scaffolds for cartilage tissue engineering. From the clinical editor: These authors have developed spatially-controlled nano-topography in the form of nano-pillar, nano-hole and nano-grill on polycaprolactone surface via thermal nanoimprinting, and the effects of chondroitin sulfate-coated nano-topographies on cell characteristics and chondrogenic differentiation of human mesenchymal stem cells (MSC) were investigated. It has been concluded that MSC differentiation is sensitive to surface nano-topography, and certain nano-imprinted surfaces are more useful than others for cartilage tissue engineering.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
DL-Proline, ReagentPlus®, 99%
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Proline, European Pharmacopoeia (EP) Reference Standard