Skip to Content
Merck
All Photos(2)

Documents

V900399

Sigma-Aldrich

L-Cysteine hydrochloride

≥98%, , Vetec

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)COOH · HCl
CAS Number:
Molecular Weight:
157.62
Beilstein:
3560277
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

product name

L-Cysteine hydrochloride, Vetec, reagent grade, ≥98%

grade

reagent grade

product line

Vetec

Assay

≥98%

SMILES string

Cl[H].N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S.ClH/c4-2(1-7)3(5)6;/h2,7H,1,4H2,(H,5,6);1H/t2-;/m0./s1

InChI key

IFQSXNOEEPCSLW-DKWTVANSSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

NMDA glutamatergic receptor agonist that is also an agonist at AMPA glutamatergic receptors at high concentrations.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabine A Tanner et al.
The British journal of nutrition, 114(5), 746-755 (2015-08-28)
Modulating the gut microbiota via dietary interventions is a common strategy to enhance the natural defence mechanisms of the host. Several in vitro studies have highlighted the probiotic potential of Bifidobacterium thermophilum RBL67 (RBL67) selected for its anti-Salmonella effects. The
Chen Ding et al.
Microbial ecology, 70(2), 433-444 (2015-03-19)
Anaerobic alkane degradation via methanogenesis has been intensively studied under mesophilic and thermophilic conditions. While there is a paucity of information on the ability and composition of anaerobic alkane-degrading microbial communities under low temperature conditions. In this study, we investigated
Joanne Mawhinney et al.
Acta odontologica Scandinavica, 73(3), 188-194 (2014-11-12)
The aim of this study was to compare internal bacterial colonization in two implant systems, one screw root form (SRF) with an external hexagon connection and one plateau root form (PRF) with a Morse taper internal connection. Thirty-two implants; 12
Min Hyung Kang et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 87(3), 489-499 (2014-04-08)
To facilitate selective drug delivery to hepsin (Hpn)-expressing cancer cells, the RIPL peptide (IPLVVPLRRRRRRRRC; 16mer; 2.1 kDa) was synthesized as a novel cell penetrating/homing peptide (CPHP) and conjugated to a liposomal carrier. RIPL peptide-conjugated liposomes (RIPL-Lipo) were prepared by conjugating
Ilaria Zanotti et al.
Applied microbiology and biotechnology, 99(16), 6813-6829 (2015-04-13)
Bifidobacteria are members of the human gut microbiota, which are known to influence the metabolic abilities of their host. Here, we investigated the capabilities of bifidobacteria to reduce cholesterol levels in synthetic growth media, clearly demonstrating assimilation of this molecule

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service