Merck
  • Evidence for cholesterol-lowering activity by Bifidobacterium bifidum PRL2010 through gut microbiota modulation.
Evidence for cholesterol-lowering activity by Bifidobacterium bifidum PRL2010 through gut microbiota modulation.
Applied microbiology and biotechnology (2015-04-13)
Ilaria Zanotti, Francesca Turroni, Antonio Piemontese, Leonardo Mancabelli, Christian Milani, Alice Viappiani, Gilda Prevedini, Borja Sanchez, Abelardo Margolles, Lisa Elviri, Bernini Franco, Douwe van Sinderen, Marco Ventura
ABSTRACT
Bifidobacteria are members of the human gut microbiota, which are known to influence the metabolic abilities of their host. Here, we investigated the capabilities of bifidobacteria to reduce cholesterol levels in synthetic growth media, clearly demonstrating assimilation of this molecule by particular bifidobacterial strains, including Bifidobacterium bifidum PRL2010 (LMG S-28692). The transcriptomic analysis of PRL2010 cells cultivated in the presence of cholesterol revealed a significantly increased transcription level of genes encoding putative transporters and reductases, indicative of specific mechanisms for cholesterol assimilation as well as cholesterol conversion to coprostanol. Cholesterol lowering activity of B. bifidum PRL2010 cells was further evaluated by means of an in vivo murine model, showing that the fecal microbiota of mice is modified toward those bacteria involved in the metabolism of cholesterol.
Product Description
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, from non-animal source, BioReagent, suitable for cell culture, ≥98.0%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, ≥98% (TLC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Benzenesulfonyl hydrazide, 98%