1369431
USP
Lomustine Related Compound B
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
1-(2-Chloroethyl)-3-cyclohexylurea, N-(2-Chloroethyl)-N′-cyclohexylurea
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
lomustine
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
InChI
1S/C9H17ClN2O/c10-6-7-11-9(13)12-8-4-2-1-3-5-8/h8H,1-7H2,(H2,11,12,13)
InChI key
BIOZXMXBLXQEBQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service